Metyrapone ≥99%
200 mg
Catalog # 89157-886
Supplier: Enzo Life Sciences
Supplier #: BML-EI256-0200
Jump to:
Product Details & Documents
Inhibits cytochrome P-450-mediated prostaglandin ω/ω-1 hydroxylase activity.
- Cytochrome P450 inhibitor
White solid.
Soluble in water (25 mg/ml), DMSO (25 mg/ml) or ethanol (25mg/ml).
Synonyms: 2-Methyl-1,2-di(pyridin-3-yl)propan-1-one
- CAS Number:54-36-4
- Formula:C₁₄H₁₄N₂O
- MDL Number:MFCD00006397
- Molecular Weight:226.34 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Size:200 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



