Order Entry
Canada
ContactUsLinkComponent
 
Sheep Cyclooxygenase (from Seminal vesicles)
Catalog # 89157-548
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Sheep Cyclooxygenase (from Seminal vesicles)
Catalog # 89157-548
Supplier:  Enzo Life Sciences
Supplier Number:  BML-EH001-0001
CAS Number:  

Specifications

  • Source:
    Seminal vesicles
  • Species:
    Sheep
  • Size:
    1 mg
  • Form:
    Liquid
  • Enzyme name:
    Cyclooxygenase
  • Enzyme activity:
    Specific activity with arachidonic acid is 24 units/mg protein. One unit will consume one micromole of oxygen per minute at 30 °C
  • Purity:
    >95%
  • Formulation:
    In 100 mM TRIS/HCl, pH 8.0, containing 1% Tween 80.
  • Cat. no.:
    89157-548
  • Supplier No.:
    EH001-0001

Specifications

About this item

Isolated from ram seminal vesicles.