Order Entry
Canada
ContactUsLinkComponent
FLUOR DE LYS®-Green HDAC Fluorometric Activity Assay Kit, Enzo Life Sciences
FLUOR DE LYS®-Green HDAC Fluorometric Activity Assay Kit, Enzo Life Sciences
Supplier:  Enzo Life Sciences
Certificates
undefined
FLUOR DE LYS®-Green HDAC Fluorometric Activity Assay Kit, Enzo Life Sciences
Supplier:  Enzo Life Sciences

Specifications

  • Storage conditions:
    –80 °C (Long Term)
  • Application:
    Fluorescent Detection, HTS Activity Assay, Cell-based Assays
  • Purity:
    ≥95% (HPLC)
  • Formulation:
    50mM solution in DMSO.

Specifications

Reagents

Product Information

Frequently Bought Together