Cathepsin S Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-494
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:Cathepsin S Drug Discovery Kit
- Size:1 Kit
- Cat. no.:89156-494
- Storage Temperature:-80 °C
- Application:Fluorescent detection, HTS
Activity assay - Supplier No.:AK431-0001
Specifications
About this item
A QuantiZyme™ Assay System
- Complete system designed to screen modulators of cathepsin S activity
- The assays are performed in a convenient 96-well microplate format
The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit.
Delivery: Kit contains Cathepsin S enzyme (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer and 1/2-volume white microplate.