Matrix Metalloproteinase-20 (MMP-20) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-490
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Colorimetric Detection, HTS Activity Assay
- Cat. no.:89156-490
- Supplier No.:AK422-0001
Specifications
About this item
A QuantiZymeTM Assay System
The MMP-20 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-20 (MMP-20, enamelysin) using a thiopeptide as a chromogenic substrate.
Delivery: Kit contains MMP-20 enzyme (human) (recombinant), substrate, a prototypic control inhibitor (NNGH3), assay buffer, 96-well microplate and a detailed instruction booklet.