Matrix Metalloproteinase-8 (MMP-8) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-478
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Colorimetric Detection, HTS
Activity Assay - Cat. no.:89156-478
- Supplier No.:AK414-0001
Specifications
About this item
A QuantiZymeTM Assay System
The MMP-8 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a thiopeptide as a chromogenic substrate.
Delivery: Kit contains MMP-8 Enzyme (human) (recombinant), Substrate,, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.