Matrix Metalloproteinase-13 (MMP-13) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-474
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 Kit
- Environmentally Preferable:
- Storage conditions:−80 °C
- Application:Colorimetric Detection, HTS
- Cat. no.:89156-474
- Supplier No.:AK412-0001
Specifications
About this item
A QuantiZymeTM Assay System
The MMP-13 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-13 (MMP-13, collagenase-3) using a thiopeptide as a chromogenic substrate.
Delivery: Kit contains MMP-13 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.