Order Entry
Canada
ContactUsLinkComponent
Matrix Metalloproteinase-9 (MMP-9) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Matrix Metalloproteinase-9 (MMP-9) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-472
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Matrix Metalloproteinase-9 (MMP-9) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Catalog # 89156-472
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK411-0001
CAS Number:  

Specifications

  • Size:
    1 Kit
  • Storage conditions:
    −80 °C
  • Application:
    Fluorescent Detection, HTS
    Activity Assay
  • Cat. no.:
    89156-472
  • Supplier No.:
    AK411-0001

Specifications

About this item

A QuantiZymeTM Assay SystemThe MMP-9 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH

Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.

Delivery: Kit contains MMP-9 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.