Matrix Metalloproteinase-7 (MMP-7) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Catalogue # 89156-464
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Fluorescent Detection, HTS
Activity Assay - Cat. no.:89156-464
- Supplier No.:AK407-0001
Spécifications
À propos de cet article
A QuantiZymeTM Assay System
The MMP-7 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-7 (MMP-7, matrilysin) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Delivery: Kit contains MMP-7 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.