Matrix Metalloproteinase-1 (MMP-1) Fluorometric Drug Discovery Kit, Enzo Life Sciences
89156-460
:
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Fluorescent Detection, HTS, Activity Assay
- Cat. no.:89156-460
- Supplier No.:AK405-0001
A QuantiZymeTM Assay System
The MMP-1 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties
: Kit contains MMP-1 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.