Order Entry
Canada
ContactUsLinkComponent
 
Matrix Metalloproteinase-12 (MMP-12) Colorimetric Drug Discovery Kit, Enzo Life Sciences
  89156-454
 :  
undefined
Matrix Metalloproteinase-12 (MMP-12) Colorimetric Drug Discovery Kit, Enzo Life Sciences
  89156-454
 :  BML-AK402-0001
 :  

 

  • Size:
    1 Kit
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric Detection, HTS, Activity Assay
  • Cat. no.:
    89156-454
  • Supplier No.:
    AK402-0001

 

 

A QuantiZymeTM Assay System

The MMP-12 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-12 (MMP-12, macrophage elastase, metalloelastase) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well plate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet.

 : Kit contains MMP-12 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well Microplate and a detailed instruction booklet.