CV-Cathepsin L Detection Kit, Enzo Life Sciences
89156-444
:
- Size:1 Kit
- Storage conditions:4 °C
- Application:Fluorescence Microscopy, Fluorescent Detection
- Cat. no.:89156-444
- Supplier No.:AK127-0001
Based on the dye cresyl violet and the cathepsin L PheArg peptide recognition sequence, this kit allows the detection of cathepsin L activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
: Kit contains CV-(FR)2 reagent, Acridine Orange, Hoechst 33342 stain.