ContactUsLinkComponent
MMP Inhibitor Profiling Kit, Colorimetric, Enzo Life Sciences
MMP Inhibitor Profiling Kit, Colorimetric, Enzo Life Sciences
Catalog # 89156-416
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
MMP Inhibitor Profiling Kit, Colorimetric, Enzo Life Sciences
Catalog # 89156-416
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK015-0001
CAS Number:  
Specifications
  • Size:
    1 Kit
  • Cat. no.:
    89156-416
  • Supplier No.:
    AK015-0001
  • No. of Tests:
    96
Specifications
About this item
A QUANTIZYME® Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a chromogenic substrate1,2 detected at 412nm. Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet.