BAFF Soluble (mouse), Detection Set, Enzo Life Sciences
Catalog # 89156-202
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:96 wells
- Storage temperature:−20 °C
- Cat. no.:89156-202
- Application:ELISA, Colorimetric Detection
- Supplier No.:54N-013-KI01
Specifications
About this item
For the quantitative determination of soluble mouse BAFF from biological fluids (serum and cell culture supernatant).
Delivery: Kit contains 1 vial standard (lyophilized) (STD), 1 vial coating antibody (COAT) and 1 vial detection antibody (DET).