SDMA ELISA Kit, Enzo Life Sciences
Catalog # 89156-154
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Primary antibody reactivity:Human
- Size:96 wells
- Storage conditions:4 °C (Kit), -20 °C (control and standard)
- Sensitivity:0.05 μmol/L (Range 0.1 - 2 µmol/L)
- Cat. no.:89156-154
- Supplier No.:850-331-KI01
- No. of Tests:80
Specifications
About this item
For the quantitative determination of symmetric dimethylarginine (SDMA) in human EDTA-plasma and serum.
Does not cross-react with ADMA, NMMA or L-arginine.