ADMA Direct (Mouse/Rat) ELISA Kit, Enzo Life Sciences
Catalog # 89156-148
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type:Direct
- Primary antibody reactivity:RatMouse
- Target protein:ADMA
- Description:ADMA Direct (Mouse/Rat) ELISA kit
- Size:96 wells
- Storage conditions:4 °C (kit), -20°C (control and standard)
- Sensitivity:0.05 µmol/L
- Cat. no.:89156-148
- Supplier No.:850-327-KI01
- No. of Tests:80
Specifications
About this item
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum
Does not cross-react with SDMA, NMMA or L-arginine.