Order Entry
Canada
ContactUsLinkComponent
Microcystins (Adda Specific) ELISA Kit, Enzo Life Sciences
Microcystins (Adda Specific) ELISA Kit, Enzo Life Sciences
Supplier:  Enzo Life Sciences
Certificates
undefined
Microcystins (Adda Specific) ELISA Kit, Enzo Life Sciences
Supplier:  Enzo Life Sciences
Certificates

Specifications

  • Storage conditions:
    4 °C (long term)
  • Sensitivity:
    0.1 ng/mL (Range 0.15 - 5 ng/mL)

Specifications

Related Information

Frequently Bought Together