Protein Carbonyl ELISA Kit, Enzo Life Sciences
89156-130
:
- Conjugate ELISA:HRP-Streptavidin
- Primary antibody reactivity:Species independent
- Description:Protein Carbonyl ELISA Kit
- Size:96 wells
- Sample type:Plasma, other body fluids, cell and tissue extracts
- Detection method:Photometric (450 nm)
- Time to Results:4.5 hrs
- Cat. no.:89156-130
- Supplier No.:850-312-KI01
- No. of Tests:80
The Protein carbonyl ELISA kit is an antibody-based detection set
- Detecting low levels of carbonylated proteins
- Validated in various sample types
- Fully quantitative results that surpass semi-quantitative Western blot analysis
- Assay Time: 4.5 hours
For quantitative determination of carbonylated protein levels in plasma, other body fluids, cell and tissue extracts.
It has higher sensitivity, lower background, is less labour-intensive and handles more samples per day as compared to colorimetric assays.
: Research Use Only