CD40L, Soluble (Human) ELISA Kit (High Sensitivity), Enzo Life Sciences
Catalog # 89156-128
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Primary antibody reactivity:Human
- Target protein:CD40L
- Description:CD40L, Soluble (Human) ELISA kit (high sensitivity)
- Size:96 Wells
- Storage conditions:4 °C (Kit), -20 °C (Control)
- Sensitivity:0.005 ng/mL (Range 0.08 - 5 ng/mL)
- Cat. no.:89156-128
- Supplier No.:850-311-KI01
- No. of Tests:80
Specifications
About this item
For the quantitative determination of soluble human CD40L (CD154) in cell culture supernatants, serum and other body fluids.
- Assay Time: 3 hours + 10 minutes