OXI-TEK TBARS Assay Kit, Enzo Life Sciences
Catalog # 89156-088
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:160 Tests
- Storage temperature:4 °C (Long Term)
- Application:Fluorescence Microscopy
- Cat. no.:89156-088
- Supplier No.:850-287-KI01
Specifications
About this item
The sensitivity of measuring thiobarbituric acid reactive substances (TBARS) has made this assay the method of choice for screening and monitoring lipid peroxidation, which is a major indication of oxidative stress.
Malondialdehyde (MDA) forms a 1:2 adduct with thiobarbituric acid; the adduct can be measured by fluorometry. Biological specimens contain a mixture of TBARS, including lipid hydroperoxides and aldehydes. In practice, TBARS are expressed in equivalent numbers to MDA.