Order Entry
Canada
Orders LinkContactUsLinkComponent
 

ApoSENSOR™ Cell Viability Assay Kit, Enzo Life Sciences

200 tests
Catalog # 89156-016
Supplier:   Enzo Life Sciences
 :  850-247-KI01
ApoSENSOR™ Cell Viability Assay Kit, Enzo Life Sciences
Catalog # 89156-016
Supplier:  Enzo Life Sciences
CAS Number:  
ApoSENSOR™ Cell Viability Assay Kit, Enzo Life Sciences
ApoSENSOR™ Cell Viability Assay Kit, Enzo Life Sciences

200 tests

CAS Number:  
Price
537,20 $
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The assay utilizes bioluminescent detection of the ATP level for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells

The assay utilizes the enzyme luciferase to catalyze the formation of light from ATP and luciferin. The light can be measured using a luminometer or Beta Counter. The assay can be fully automatic for high throughput (10 seconds/sample) and is highly sensitive (detects 10-100 mammalian cells/well).

 : Kit includes Nucleotide Releasing Buffer, ATP Monitoring Enzyme, Enzyme Reconstitution Buffer and ATP.

Specifications

Specifications

  • Size:
    200 tests
  • Cat. No.:
    89156-016
  • Storage conditions:
    –20 °C (Long Term)
  • Application:
    Chemiluminescence

Specifications