Cu/Zn-Superoxide Dismutase ELISA Kit, Enzo Life Sciences
89155-894
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Cu/Zn-Superoxide Dismutase ELISA kit
- Size:96 well (~ 80 tests)
- Storage conditions:4 °C (long term)
- Time to Results:< 2 hours
- Sensitivity:0.04 ng/mL (Range 0.08 to 5 ng/mL)
- Regulatory status:RUO
- Immunogen:Cu/ZnSOD
- Cat. no.:89155-894
- Supplier No.:850-033-KI01
For the quantitative determination of human Cu/Zn SOD in cell culture supernatants, serum and other body fluids.
- Assay Time: <2 hours
Late stages of apoptosis are characterized by the loss of membrane integrity and a release of the intracellular Cu/Zn SOD.