Phosphotyrosine detection set, Enzo Life Sciences
Catalog # 89155-886
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:Phosphotyrosine Detection Set
- Size:1 Set
- Cat. no.:89155-886
- Supplier No.:850-025-KI01
Specifications
About this item
This kit offers a set of seven different MAbs specific for tyrosine phosphorylation sites. Phosphorylation patterns in a given cell extract may differ when probed with different antibodies due to sequence specificity. Recognition therefore depends on two criteria: Phosphorylation and surrounding aa motif. If one of the two criteria is not fulfilled, the antibody will not detect the phosphorylation site. Specificity has been determined by epitope mapping using degenerated phosphopeptide libraries.
Ordering information: Contents: Contains 25µg each of the following MAbs:
Clone 2C8
Clone 3B12
Clone 9H8
Clone 16F4 (Prod. No. ALX-804-174)
Clone 9F1 (Prod. No. ALX-804-669)
Clone 2A5 (Prod. No. ALX-804-670)
Clone 1F9 (Prod. No. ALX-804-671)
Clone 2C8
Clone 3B12
Clone 9H8
Clone 16F4 (Prod. No. ALX-804-174)
Clone 9F1 (Prod. No. ALX-804-669)
Clone 2A5 (Prod. No. ALX-804-670)
Clone 1F9 (Prod. No. ALX-804-671)