Iberiotoxin
Catalog #: 89152-954
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:100 µg
- Reconstitution instructions:Do not remove the cap. Inject 260μl distilled water into the vial using a calibrated syringe. Dissolve the contents thouroughly.
- Protein/peptide name:Iberiotoxin
- Purity:>94% (HPLC)
- Concentration:0.1mM after reconstitution.
- Formulation:Lyophilized from aqueous acetic acid solution.
- Cat. no.:89152-954
- Supplier No.:630-058-C100
Specifications
About this item
Ca2+-activated K+ channel blocker
Potent and selective blocker of the high conductance Ca2+-activated K+ channels.
Ordering information: Quantity: 0.11mg (25nmol)
Customers Who Bought This Also Bought