To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:40 µg
- Reconstitution instructions:Reconstitute in 0.5ml of distilled cold water. Stir very gently. Do not shake or vortex. Add 0.5 ml glycerol. Stock solution received contains 50% glycerol and 300nM α-latrotoxin.
- Protein/peptide name:Latrotoxin
- Purity:>97%
- Formulation:Lyophilized
- Purification:Migrates as one distinct band when run on a 5-25% gradient polyacrylamide gel according to the method of Laemmli (Nature 227, 680 (1970)).
- Cat. no.:89152-926
- Supplier No.:630-027-C040
Specifications
About this item
Neurotoxin.
Causes massive neurotransmitter release from a wide variety of central and peripheral synaptic junctions of vertebrates using Ca2+-dependent and Ca2+-independent pathways. A useful pharmacological tool in the studies of synaptic vesicles exocytosis of different neutrotransmitters.
Isolated from Latrodectus tredecimguttatus.