Mouse Recombinant Netrin 1
Catalog # 89151-924
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Size:10 µg
- Endotoxin content:<0.1EU/µg purified protein (LAL test; Bio Whittaker).
- Reconstitution instructions:Reconstitute with 100µl sterile distilled water. Resuspended protein should be used immediately upon reconstitution to avoid loss in activity due to non-specific binding to the vial. Further dilutions should be made with medium containing 5% fetal calf se
- Protein/peptide name:Netrin 1
- Purity:>95% (SDS-PAGE)
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Cat. no.:89151-924
- Supplier No.:522-124-2010
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of mouse netrin-1 (aa 27-604) is fused at the C-terminus to a FLAG®-tag.