Order Entry
ContactUsLinkComponent
Mouse Recombinant Adiponectin
Mouse Recombinant Adiponectin
Catalog #: 89151-802
Supplier:  Enzo Life Sciences
CAS Number:  
ADIPONECTIN MOUSE REC
Mouse Recombinant Adiponectin
Catalog #: 89151-802
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    50 µg
  • Endotoxin content:
    \<0.1EU/µg purified protein (LAL test; Bio Whittaker).
  • Biological activity:
    Inhibits serum glucose production in mice. Protects mouse hepatocytes from TNF-α-induced cell death in vitro.
  • Reconstitution instructions:
    Reconstitute with 100µl sterile water.
  • Protein synonyms:
    ACDC
  • Protein/peptide name:
    Adiponectin
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0.5mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Cat. no.:
    89151-802
  • Supplier No.:
    522-059-C050

Specifications

About this item

Produced in HEK 293 cells. Mouse adiponectin (aa 18-247) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.

Mimics serum adiponectin by forming high molecular weight (HMW) and hexameric species.

Customers Who Bought This Also Bought