Human Recombinant BAFF (soluble) (from E. coli)
Catalog #: 89151-730
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 µg
- Endotoxin content:\<0.1EU/µg purified protein (LAL test; Bio Whittaker).
- Biological activity:Mediates splenocyte survival. Induces monocyte survival.
- Reconstitution instructions:Reconstitute with 100µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:BAFF (soluble)
- Purity:>95% (SDS-PAGE)
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Cat. no.:89151-730
- Supplier No.:522-025-C010
Specifications
About this item
Produced in E. coli. The extracellular domain of human BAFF (aa 83-285) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.
Customers Who Bought This Also Bought