Human Recombinant CD40L (soluble) (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.