Order Entry
Canada
ContactUsLinkComponent
Human Recombinant TNF alpha (soluble) (from E. coli)
Human Recombinant TNF alpha (soluble) (from E. coli)
  89151-692
 :  
undefined
Human Recombinant TNF alpha (soluble) (from E. coli)
  89151-692
 :  522-008-C050
 :  

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Endotoxin content:
    <0.1EU/µg purified protein (LAL test; Bio Whittaker).
  • Biological activity:
    Exerts its biological activity in a concentration range of 0.1-1ng/ml (WEHI 164 cells).
    ED50: 0.1ng/ml (WEHI 164 cells)
  • Reconstitution instructions:
    Reconstitute with 50µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    TNF alpha (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Cat. no.:
    89151-692
  • Supplier No.:
    522-008-C050

 

 

Produced in E. coli. The extracellular domain of human TNF-α (tumor necrosis factor-α) (aa 85-223) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.