Order Entry
ContactUsLinkComponent
Human Recombinant IL1B (from E. coli)
Human Recombinant IL1B (from E. coli)
Catalog #: 89151-652
Supplier:  Enzo Life Sciences
CAS Number:  
IL 1BETA HUMAN RECOMBINAN 10MG
Human Recombinant IL1B (from E. coli)
Catalog #: 89151-652
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 µg
  • Endotoxin content:
    \<1EU/µg protein.
  • Biological activity:
    Exerts in vitro biological activity in the range of 0.1-10ng/ml.
    ≥1x107U/mg protein. Determined by proliferation assay using human TF-1 (T. Kitamura, et al.; Int. Immunol. 3, 571 (1991)). The ED50 for this effect is typically 0.2-2ng/ml. Results were compared with the international IL-1 reference standard preparation (
  • Reconstitution instructions:
    Reconstitute with water, PBS or aqueous buffer to a concentration of 100µg/ml. For further dilution carrier protein (~1%) should be added to avoid loss of bioactivity.
  • Protein synonyms:
    IL-1beta
  • Protein/peptide name:
    IL1B
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Lyophilized. Salt-free.
  • Grade:
    Cell culture Grade
  • Cat. no.:
    89151-652
  • Supplier No.:
    520-001-C010

Specifications

About this item

Potent immunomodulator. Produced in E. coli.

Potent immunomodulator. Mediates a wide range of immune and inflammatory responses including the activation of B and T cells.

Customers Who Bought This Also Bought