ContactUsLinkComponent
PBS (Endotoxin-Free), Enzo Life Sciences
PBS (Endotoxin-Free), Enzo Life Sciences
Catalog # 89151-648
Supplier:  Enzo Life Sciences
CAS Number:  
PBS (Endotoxin-Free), Enzo Life Sciences
Catalog # 89151-648
Supplier:  Enzo Life Sciences
Supplier Number:  505-007-LD15
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PBS Endotoxin content <0.0002 EU/µg (LAL test; BioWhittaker)
  • Size:
    1.5 ml
  • Cat. no.:
    89151-648
  • Supplier No.:
    505-007-LD15
Specifications
About this item
Sterile and endotoxin-free PBS for use with TLRgrade™ or endotoxin-free grade reagents.
Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.