BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:387334-31-8
:
Formula:C₁₀H₁₇NO₃
MDL Number:MFCD32196548
:199.3 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity (1H-NMR, EPR):≥99%
Specification Test Results
Frequently Bought Together