
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98% (by non aqueous titration)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:18390-00-6
:
Formula:C₁₃H₁₇N₂O₂
MDL Number:MFCD00059929
:233.29 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (Nonaqueous Titration)
- Purity Detail:Purified by nonaqueous titration
Specification Test Results
Frequently Bought Together