Order Entry
Canada
ContactUsLinkComponent
Hygromycin B 100 mg/mL in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Hygromycin B 100 mg/mL in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Catalog # 89150-432
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Hygromycin B 100 mg/mL in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Catalog # 89150-432
Supplier:  Enzo Life Sciences
Supplier Number:  380-309-G001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 g
  • Environmentally Preferable:
  • Cat. no.:
    89150-432
  • Supplier No.:
    380-309-G001

Specifications

About this item

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
 :
 :527.5 g/mol
Storage Temperature:Refrigerator
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479