To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Source:Bacillus colistinus
- CAS No.:1066-17-7
1264-72-8 - Size:500 mg
- Storage conditions:Frezzer
- MDL No.:MFCD00146495
- Protein synonyms:Polymyxin E sulfate salt
- Protein/peptide name:Colistin sulphate
- Molecular formula:C₁₀₄H₂₀₆N₃₂O₄₆S₅
- Cat. no.:89150-322
- Supplier No.:380-272-M500
Specifications
About this item
Polymyxin antibiotic
Mixture of >30 compounds, primarily colistin A and B
Binds to lipids on the cell cytoplasmic membrane. Disrupts cell wall integrity. Effective against Gram-negative bacteria.
Soluble in water (50 mg/mL)