To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:50 µg
- Reconstitution instructions:Reconstitute in 10 mM HEPES buffer, pH7.2. (We recommend adding a carrier protein such as BSA to the reconstitution buffer and any subsequent dilution buffers to prevent absorbtion on plastic surfaces.)
- Protein/peptide name:Convulxin
- Purity:>90% (SDS-PAGE)
- Formulation:Lyophilized
- Cat. no.:89149-060
- Supplier No.:350-100-C050
Specifications
About this item
Heterodimeric C-type lectin
Isolated from Crotalus durissus terrificus snake venom.
Heterodimeric C-type lectin. Activates mammalian platelets via binding and clustering of p62/GPVI-receptors under physiological conditions.