Z-VEID-FMK (Ready-to-Use), Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase-6 inhibitor
Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.