Z-DEVD-Fmk
Supplier: Enzo Life Sciences
Certificates
About this item
Inhibitor of caspase-3 and -7
Irreversible, cell permeable inhibitor of caspase-3 and -7.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.