AC-LEVD-AFC
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase-4 substrate
Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (Prod. No. ALX-260-083); the AFC fluorophore is more sensitive.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.