Ac-VDVAD-AMC
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase-2 inhibitor
Fluorogenic substrate for caspase-2. Preferred cleavage site for caspase-2 (Km=80μM).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.