Z-YVAD-pNA, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase-1 substrate
Chromogenic substrate for caspase-1 with increased cell permeability. λmax of pNA is 400nm.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.