AC-IETD-AMC
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase and granzyme substrate
Fluorogenic substrate for caspase-6 and -8 and granzyme B. Also a substrate for caspase-10.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.