Ac-YVAD-CMK
Supplier: Enzo Life Sciences
Certificates
About this item
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Inhibits Fas-mediated apoptosis.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.