Z-VAD-FMK, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase inhibitor
Irreversible, cell permeable broad-spectrum caspase inhibitor.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.