Human Recombinant Annexin V (from E. coli), FITC (Fluorescein Isothiocyanate)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm; Em(max): 530 nm).
Ordering information: Quantity: 20 tests/100µl; 100 tests/500µl; 300 tests/1'500µl.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.