Human Recombinant Matriptase 2
125 U
Catalog # 89144-978
Supplier: Enzo Life Sciences
Supplier #: ALX-201-752-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag.
- Trypsin-like serine protease.
Matriptase-2 is a potent negative regulator of hepcidin.
Specifications
Specifications
- Size:125 U
- Cat. No.:89144-978
- Enzyme type:Recombinant
- Enzyme activity:~2.5 U/µl. One unit is defined at the amount of enzyme that releases 1 nmol of AMC per min. at 37 °C in the presence of 50 µM Boc-Gln-Ala-Arg-AMC.
- Form:Liquid
- Formulation:In 50 mM HEPES, pH 9.0, containing 400 mM sodium chloride and 20% glycerol.
- Concentration:100 nM
- Species:Human
- Enzyme name:Matriptase 2



