Human Recombinant CARM1 (from Insect cells (Sf9)), HIS Tag
Catalog #: 89144-426
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Size:2 µg
- Tag sequence:HIS Tag
- Biological activity:~50-100ng are required for standard HAT assay.
- Protein/peptide name:CARM1
- Purity:>90% (SDS-PAGE)
- Concentration:0.5mg/ml
- Formulation:Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50mM imidazole and 10% glycerol.
- Cat. no.:89144-426
- Supplier No.:201-283-C002
Specifications
About this item
Produced in SF21 cells. Fused to a His-tag.
Customers Who Bought This Also Bought