To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Size:100 µg
- Biological activity:Inhibits human leukocyte elastase (Ki=0.17nM) and human proteinase 3 (Ki=0.42nM) .
Inhibition of human leukocyte elastase: ≥90% - Reconstitution instructions:Reconstitute in sterile distilled water. Further dilutions should be made in aqueous buffers, pH 4.0-8.0.
- Protein/peptide name:Elafin
- Purity:>90% (HPLC, SDS-PAGE, MS)
- Formulation:Lyophilized
- Cat. no.:89144-350
- Supplier No.:201-240-C100
Specifications
About this item
Produced in yeast.
Belongs to the chelonianin family of protease inhibitors and is involved in the physiological control of neutrophil serine proteinases, where it targets NE and proteinase 3. Shows antiprotease, antibacterial and antiinflammatory activity and primes innate immunity.