Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Human Recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Catalog # 89144-214
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Catalog # 89144-214
Supplier:  Enzo Life Sciences
Supplier Number:  201-177-C010
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Size:
    10 µg
  • Form:
    Liquid
  • Enzyme name:
    Guanylyl cyclase alpha1beta1 (soluble)
  • Enzyme activity:
    - Basal: ≥0.075 µmol/mg/min.
    - 100 µM DEA NONOate (Prod. No. ALX-430-034) ≥21.6 µmol/mg/min.
    - 1 µM BAY 41-2272 (Prod. No. ALX-420-030): ≥1.2 µmol/mg/min.
  • Purity:
    ≥90% (SDS-PAGE)
  • Concentration:
    0.4 mg/ml
  • Formulation:
    In 40 mM TEA, pH 7.4 containing 2 mM DTT, 1 mM MgCl₂  and 30% glycerol.
  • Cat. no.:
    89144-214
  • Supplier No.:
    201-177-C010

Specifications

About this item

Produced in Sf9 cells. Fused at the C-terminus to a His-tag.